Iron, Plasma Standard Solution, Specpure®, Fe 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Iron, plasma standard solution, Specpure|r, Fe 10,000^mg/ml
Molecular Formula: (Fe(NO3)3).9H2O in 5% HNO3
Physical Form: Liquid
Odor: Acidic
UN No.: 3265
MDL No.: MFCD00010999
Specifications
- MDL No.:MFCD00010999
Specifications
Related Information
Frequently Bought Together