Fluid Impervious PE, Over-the-Head Thumb-Hook Gown, Tronex®
Supplier: Tronex
Certificates
About this item
Constructed from embossed polyethylene (PE) to provide an effective fluid barrier, these gowns comfortably protect wearers who need a high level of fluid protection
- Thumb-hook cuff designed for secure fit and full arm coverage when worn under gloves
- Superior durability and versatility with high tensile strength
- Provides full frontal coverage and an open back for ventilation
- Heat-sealed seams offer enhanced fluid protection
- Apron-style neck opening and perforated back shoulder allow for easy donning and pull-away removal
- Waist ties provide adjustability for a contoured fit
A gown featuring extra-long ties is also available for added versatility.
Specifications
- Catalog No:
- 10152-936
- Garment application:
- Medical, Clinical