ContactUsLinkComponent
Foam Biopsy Pads, Globe Scientific
Foam Biopsy Pads, Globe Scientific
Catalog # 10022-166
Supplier:  Globe Scientific
undefined
Foam Biopsy Pads, Globe Scientific
Catalog # 10022-166
Supplier:  Globe Scientific
Supplier Number:  190195
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Color:
    Blue
  • Description:
    Foam Biopsy Pads
  • Cat. No.:
    10022-166
  • Supplier no.:
    190195
Specifications
About this item
The Globe Scientific foam biopsy pads are used to hold biopsies in place and prevent their loss during processing
They are produced from a special polyester urethane foam, which is verified for consistency throughout in order to ensure optimum solvent flow. Simply sandwich the biopsy sample between two pads and insert into a cassette for processing.