ContactUsLinkComponent
LEADING LIGHT® Wnt Reporter Assay Starter Kit, Enzo Life Sciences
LEADING LIGHT® Wnt Reporter Assay Starter Kit, Enzo Life Sciences
Catalog # 10014-514
Supplier:  Enzo Life Sciences
CAS Number:  
LEADING LIGHT® Wnt Reporter Assay Starter Kit, Enzo Life Sciences
Catalog # 10014-514
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-61001-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    10014-514
  • No. of wells:
    96
  • Supplier no.:
    ENZ-61001-0001
Specifications
About this item
Chemiluminescent cell-based assay, for discovering Wnt pathway modulators
The LEADING LIGHT® Wnt Reporter Assay Starter Kit is a cell-based luciferase activity test suitable for 96 well plate format. The system contains an engineered 3T3 mouse fibroblast cell line, which expresses the firefly luciferase reporter gene under the control of Wnt-responsive promoters (TCF/LEF). The luciferase activity from the reporter gene in this cell line can be up-regulated in a dose-dependent manner upon the addition of exogenous Wnt protein/Wnt agonist or down-regulated by a further addition of a Wnt antagonist to the cell culture medium. This system can be used to elucidate the functions/activities of different Wnt-related ligands such as Wnt, DKK, etc. This system can also be used for screening small molecules and antibodies for their ability to act as Wnt inhibitors or Wnt agonists.The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.
Ordering information: Contains LEADING LIGHT® Wnt reporter cell line, cell assay and growth medium concentrate, cell freezing medium, luciferase substrate and assay buffer, HEPES, Wnt3a protein (positive control) and Dkk-1 protein.