Order Entry
United States
ContactUsLinkComponent
BIOARRAY™ Low Input RNA Amplification and Biotin Labeling System, Enzo Life Sciences
BIOARRAY™ Low Input RNA Amplification and Biotin Labeling System, Enzo Life Sciences
Catalog # 10014-574
Supplier:  Enzo Life Sciences
CAS Number:  
BIOARRAY™ Low Input RNA Amplification and Biotin Labeling System, Enzo Life Sciences
Catalog # 10014-574
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-42422-0010
CAS Number:  

Specifications

  • Description:
    Low Input RNA Amplification and Biotin Labeling System BIOARRAY™
  • Cat. No.:
    10014-574
  • No. of reactions:
    10
  • Supplier no.:
    ENZ-42422-0010

Specifications

About this item

Lower RNA Input with High Yield – High Quality Biotin-labeling

The BIOARRAY™ Low Input RNA Amplification with Biotin Labeling System provides an optimized protocol and reagents for the production of biotin-labeled antisense RNA (aRNA) from total cellular RNA samples in less than 24 hours for gene expression analysis.

The BIOARRAY™ Low Input RNA Amplification with Biotin Labeling System provides an optimized protocol and reagents for the production of biotin-labeled antisense RNA (aRNA) from total cellular RNA samples in less than 24 hours for array analysis. This kit has been improved to yield sufficient aRNA quantity for most microarray platforms (10 µg minimum) with total RNA input of as low as 20 ng and up to 500 ng. Entire amplification reaction can be performed in a single tube and the biotin-labeled aRNA can be purified using either magnetic beads or purification columns. The BIOARRAY™ Low Input aRNA Amplification and Biotin Labeling System has been optimized for superior performance with reduced variability and improved reproducibility and data quality. The complete system is composed of reagents for cDNA synthesis and in vitro transcript labeling.