Specifications
- Description:Fluid dispensing connector for filling unit-dose syringes, Sterile
- Cat. No.:10017-726
- Supplier no.:415080
Specifications
About this item
Fluid dispensing connector for filling unit-dose syringes from a large master syringe.
- Provides a secure connection of two syringes
- Luer-lock connections on both sides
- Easy air elimination from unit-dose syringe
- Large grip-plates
Easy-to-use and minimizes contamination. Air is always located in the upper end of the master syringe.