Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with MAS™ Omni-CORE™ Controls
Three distinct levels; Liquid, ready-to-use; Human serum based matrix; Assayed values available for most analyzers.
Analytes: Acetaminophen, Albumin, Alkaline Phosphatase, alpha-1-Acid Glycoprotein, alpha-1-Antitrypsin, alpha-2-Macroglobulin, ALT, Amikacin, Amylase, Amylase (pancreatic), Antistreptolysin O (ASO), Apolipoprotein A1, Apolipoprotein B, AST, Beta 2 Microglobulin, Bile Acids, Bilirubin Conjugated, Bilirubin Direct (DBIL), Bilirubin Neonatal, Bilirubin Total (TBIL), Bilirubin Unconjugated, BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Ceruloplasmin, Chloride, Cholesterol, CK, CO2, Copper, Cortisol, C-Reactive Protein (CRP), Creatinine, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, Gentamicin, GGT, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HDL Cholesterol, Immunoglobulin A (IgA), Immunoglobulin E (IgE), Immunoglobulin G (IgG), Immunoglobulin M (IgM), Iron, Iron Binding Capacity (IBC), Iron Binding Capacity, Total (TIBC), Iron Binding Capacity, Unsaturated (UIBC), Lactic Acid, LDH, LDL-Cholesterol, Lidocaine, Lipase, Lipoprotein (a), Lithium, Magnesium, Methotrexate, NAPA, Osmolality, Phenobarbital, Phenytoin, Phospholipids, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Protein Electrophoresis, Pseudocholinesterase (PCHE), Quinidine, Rheumatoid Factor, Salicylate, Sodium, T4 Total (thyroxine), Theophylline, Tobramycin, Total Protein, Transferrin, Tricyclic Antidepressant, Triglycerides, TSH (thyroid-stimulating hormone), T-Uptake, Uric Acid, Valproic Acid, Vancomycin, Zinc (refer to package insert for specific analyte and stability claims).
Value assignment is provided for key integrated instrument systems including Abbott Architect, Beckman Coulter AU, Synchron and UniCel, Ortho-Clinical Diagnostics VITROS, Roche Cobas and Elecsys systems, Siemens Advia, Dimension and Dimension Vista. Use Thermo Scientific LabLink xL QAP web based program to track QC.