About this item
Ensure accurate results in testing of traditional cardiac marker panels as well as the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase, and Procalcitonin with the MAS™ Omni-CARDIO™ Control.
Matrix: Human Serum; Packaging: 6 x 3mL assayed; Value assignment provided for key instrument systems.
Analytes: Beta Human Chorionic Gonadotropin (b-hCG), Brain Natriuretic Peptide-32 (BNP-32), Creatinine Kinase-MB (CK-MB), D-Dimer, Digitoxin, High Sensitivity C-Reactive Protein (hsCRP), Human Chorionic Gonadotropin (hCG), Myeloperoxidase (MPO), Myoglobin, N-Terminal Brain Natriuretic Peptide (NT-proBNP), Procalcitonin (PCT), Total Creatinine Kinase (Total CK), Troponin I, Troponin (refer to package insert for specific analyte and stability claims).