Puritan® DNA-Free PurFlock® Ultra Tipped Applicator with Transport Tube, Puritan Medical Products
Catalog # 89238-146
Supplier: Puritan Medical Products
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Description:DNA-Free Standard Flock Swab with Plastic Handle and Tube
- Cat. No.:89238-146
- Tip diameter:5 mm (0.2")
- Tip style:Standard Profile
- Overall length:153 mm (6.04")
- Supplier no.:253306UTTFDNA
Specifications
About this item
PurFlock® Ultra's uniquely designed fiber structure accounts for better cell yield in the specimen, particularly where sample size is limited
PurFlock® swab purity ensure ideal use for PCR, molecular assays, DNA testing, and forensic applications, as well as direct antigen testing.
Tested by an independent lab to the most stringent standards (less than 25pg of human DNA). Certified to be DNA-free, DNase-free, and RNase-free.