ContactUsLinkComponent
Whizard® Z40 Armguard, Wells Lamont
Whizard® Z40 Armguard, Wells Lamont
Catalog # 80090-819
Supplier:  Wells Lamont
Product available on GSA Advantage®
undefined
Whizard® Z40 Armguard, Wells Lamont
Catalog # 80090-819
Supplier:  Wells Lamont
Supplier Number:  135364
Product available on GSA Advantage®
Specifications
  • Type:
    Wide fit
  • Garment application:
    Industrial,
    Laboratory
  • Risk area:
    Cuts
  • Color:
    Blue
  • Cat. No.:
    80090-819
  • Supplier no.:
    135364
Specifications
About this item
Constructed from a combination of high performance materials, Z40 armguards offer ANSI level A6 cut protection.
  • Unique Knit Structure Allows Breathing
  • Wide Fit (22")
  • Reversible for extended wear
  • Clip Provided
Its unique knit structure allows breathing, and the provided clip allows users to attach the sleeve to clothing.
Recommended industries: Steel processing, metal handling, food processing
Certifications: ANSI Cut Level A6.
Ordering information: This armguard is sold in multiples of six pieces.