Uni-Dose® Vials, Type III, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These vials are manufactured from Wheaton 900 soda-lime amber glass for use with Wheaton tamper evident safety cap.
Large labeling surface for easy product identification.
Use the 224316 crimper to secure the tamper-evident safety caps (Cat. No. W224198 or W224199) to the bottle or vial.
Certifications: Conforms to USP Type III requirements.
Safety Caps
Product Information
Frequently Bought Together