Order Entry
United States
ContactUsLinkComponent
POLYVIEW® (Mouse on Mouse-HRP) IHC Kit, Enzo Life Sciences
POLYVIEW® (Mouse on Mouse-HRP) IHC Kit, Enzo Life Sciences
Catalog # 10014-544
Supplier:  Enzo Life Sciences
CAS Number:  
POLYVIEW® (Mouse on Mouse-HRP) IHC Kit, Enzo Life Sciences
Catalog # 10014-544
Supplier:  Enzo Life Sciences
Supplier Number:  950-114-0100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    POLYVIEW® (mouse on mouse-HRP) IHC kit
  • Storage Temperature:
    + 4°C
  • Processing Time:
    ~2 hours
  • Cat. No.:
    10014-544
  • Supplier no.:
    950-114-0100
  • No. of tests:
    100

Specifications

About this item

Highest sensitivity, lowest background nanopolymer detection reagent for use with HIGHDEF® chromogens.

Ordering information: Contains IHC tissue primer, blocking reagent, anti-mouse HRP polymer, DAB buffer, DAB chromogen, empty mixing bottle for DAB.
Caution: RUO - Research Use Only