ContactUsLinkComponent
Tactile Monofilament Evaluator, Foot Set
Tactile Monofilament Evaluator, Foot Set
Catalog # 470119-636
undefined
Tactile Monofilament Evaluator, Foot Set
Catalog # 470119-636
Supplier Number:  3005262
Specifications
  • Description:
    Tactile Monofilament Evaluator, Foot Set, 6 Pieces
  • Cat. No.:
    470119-636
  • Supplier no.:
    W54066
Specifications
About this item
The baseline tactile monofilament evaluator, foot set, measures cutaneous sensory perception threshold of the feet. Used to test sensation levels, and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility.
Features 2.83, 3.61, 4.31, 4.56, 5.07 and 6.65 sized monofilaments.