ContactUsLinkComponent
The Vision™ Headlight, Emergency Medical International
  10756-144
undefined
The Vision™ Headlight, Emergency Medical International
  10756-144
 :  204
  • Description:
    Head Lamp, Yellow
  • Cat. No.:
    10756-144
  • Supplier no.:
    204
The Vision™ with its 60 lumens of LED light gives you hands free lighting instantly
The brilliant 3 watt bulb gives the responder a choice of high beam, low beam, or strobe. The lightweight headlamp is made of an aluminum alloy and ABS plastic and has an easy to operate side switch even when wearing gloves. The Vision™ has an adjustable elastic headband and a 90° tilting multi-position head. It’s great for search and rescue teams and can be used in all weather conditions. It has 100,00 hours of LED light and takes 3 AAA batteries.