Specifications
- Antibody Type:Primary
- Antigen Name:Bone marrow stromal cell antigen 2
- Antigen Symbol:BST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Format:IgG
- Form:Liquid (sterile filtered)
- Gene ID:69550
- Antigen Synonyms:2310015I10Rik|CD317|Bst-2|GREG|DAMP-1|C87040
- Concentration:1 mg/mL by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-Bst2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 18 amino acid synthetic peptide near the N-terminus of the mouse Bst2.
- Purification:Anti-Bst2 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with Bst2 from other sources has not been determined.
- Cat. No.:10800-482
- Supplier no.:600-401-Z95
Specifications
About this item
Bst2 was first identified as a novel terminal B-cell-restricted antigen termed HM1.24 that is expressed on certain bone marrow cells as well as other tissues. The surface expression of Bst2 on fibroblast cell lines facilitated the stromal cell-dependent growth of DW34, a pre-B-cell line, suggesting that this protein is involved in pre-B-cell growth. Later experiments also showed it to be upregulated in several myeloma cells. More recently, Bst2 was identified an inhibitor of retroviral release from human cells whose activity is antagonized by the HIV-1 accessory protein Vpu. While Bst2 caused retention of virions on cell surfaces or endocytosis into Bst2-positive compartments, its depletion abolished the viral requirement for Vpu for virus release. This activity may represent a potential therapeutic strategy for the treatment of HIV/AIDS.
Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide
Type: Primary
Antigen: BST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse