Order Entry
United States
ContactUsLinkComponent
Anti-DFFB Rabbit Polyclonal Antibody
Anti-DFFB Rabbit Polyclonal Antibody
Catalog # 10802-446
undefined
Anti-DFFB Rabbit Polyclonal Antibody
Catalog # 10802-446
Supplier Number:  600-401-AV3

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    DNA fragmentation factor, beta subunit
  • Antigen Symbol:
    DFFB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Center
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    13368
  • Antigen Synonyms:
    5730477D02Rik|CPAN|Didff|DFF40|CAD|40kDa
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-DFF40 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 16 amino acid peptide near the internal region of human DFF40.
  • Purification:
    Anti-DFF40 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with DFF40 from other sources has not been determined.
  • Cat. No.:
    10802-446
  • Supplier no.:
    600-401-AV3

Specifications

About this item

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD for caspase activated deoxyribonuclease. The human homologue of mouse CAD was more recently identified by three groups independently and termed CPAN, DFF40, and human CAD, respectively. DFF45/ICAD is the inhibitory protein of DFF40/CAD and forms complex with DFF40/CAD. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually causes the degradation of DNA in the nuclei. Activation of DFF40/CAD, which causes DNA degradation, is the hallmark of apoptotic cell death.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Western Blot: 0.5-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat