Specifications
- Antibody Type:Primary
- Antigen Name:C1q And tumor necrosis factor related protein 7
- Antigen Symbol:C1QTNF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Format:IgG
- Form:Liquid (sterile filtered)
- Gene ID:109323
- Antigen Synonyms:8430425G24Rik|Ctrp7|5530401N20Rik
- Concentration:1 mg/mL by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-CTRP7 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 15 amino acid synthetic peptide from near the N-terminus of human CTRP7.
- Purification:Anti-CTRP7 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. These proteins often migrate in SDS-PAGE at positions other than their predicted size.
- Cat. No.:10800-954
- Supplier no.:600-401-AQ7
Specifications
About this item
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Like the other members of the adiponectin and CTRP protein family, the mature CTRP7 is secreted and can be found in the organism's circulatory system.
Recommended Dilutions: ELISA: 1:10,000-1:40,000; Western Blot: 1-4 ?g/mL; contains 0.02% (w/v) Sodium Azide
Type: Primary
Antigen: C1QTNF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat