- Antibody Type:Primary
- Antigen Name:Cyclin A2
- Antigen Symbol:CCNA2
- Clonality:Monoclonal
- Clone:CY-28
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:100 µg
- Format:IgG2a
- Form:Lyophilized
- Gene ID:890
- Antigen Synonyms:CCN1|CCNA
- Concentration:1.0 mg/mL by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Cyclin A antibody was produced in mice by repeated immunizations with a recombinant protein corresponding to bovine cyclin A.
- Purification:Anti-Cyclin A antibody was purified from mouse ascites by protein A chromatography. This product reacts with human, mouse, and bovine. Cross reactivity with Cyclin A from other sources has not been determined.
- Cat. No.:10801-044
- Supplier no.:200-301-P30
The mammalian A-type cyclin family consists of 2 members, cyclin A1 (CCNA1) and cyclin A2 (CCNA2). Human cyclin A gene is assigned to 4q26-q27. Cyclin A protein is synthesized and localized into the nucleus at the onset of S phase in nontransformed mammalian fibroblasts. This antibody is suitable for researchers interested in cancer research, apoptosis, autoimmune disease, neurodegenerative disease, cardiovascular disease, MAP Kinase Signaling, AKT Signaling, Inflammatory Signaling, and Cell Cycle research.
Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide
Type: Primary
Antigen: CCNA2
Clonality: Monoclonal
Clone: CY-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine