Anti-CD3e, CD4, CD8a Monoclonal Antibody (PE (Phycoerythrin)-Cy7®, PE (Phycoerythrin), FITC (Fluorescein Isothiocyanate)) [clone: 145-2C11, RM4-5, 53-6.7]
Catalog # 10002-340
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3e, CD4, CD8a
- Clonality:Monoclonal
- Clone:145-2C11, RM4-5, 53-6.7
- Conjugation:PE (Phycoerythrin)-Cy7®/PE (Phycoerythrin)/FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Multiple
- Reactivity:Mouse
- Size:100 µg
- Cross Adsorption:No
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Cat. No.:10002-340
- Supplier no.:91-0000-U100
Specifications
About this item
The Mouse T Lymphocyte Subset backbone panel consists of 3 anti-mouse antibodies, each conjugated to a different fluorochrome. This panel can serve as a backbone for identifying major T cell subsets, and leaves room for additional drop-in markers, if desired. An APC version of this panel is available, where the anti-CD8a antibody is conjugated to APC. Hosts: Armenian Hamster, Rat
Type: Primary
Antigen: CD3e, CD4, CD8a
Clonality: Monoclonal
Clone: 145-2C11, RM4-5, 53-6.7
Conjugation: PE (Phycoerythrin)-Cy7®, PE (Phycoerythrin), FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Multiple
Isotype:
Reactivity: