Order Entry
United States
ContactUsLinkComponent
Anti-MME Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SN5c]
Anti-MME Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SN5c]
Supplier:  Tonbo Biosciences
Certificates
Anti-MME Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SN5c]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10002-318
  • 10002-316
  • 10002-314
  • Antigen Symbol:
  • MME
  • MME
  • MME
  • Antigen Name:
  • Membrane Metallo-Endopeptidase
  • Membrane Metallo-Endopeptidase
  • Membrane Metallo-Endopeptidase
  • Antigen Synonyms:
  • NEP|CD10|CALLA|SFE
  • NEP|CD10|CALLA|SFE
  • NEP|CD10|CALLA|SFE
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • SN5c
  • SN5c
  • SN5c
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 4311
  • 4311
  • 4311
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Cross Adsorption:
  • No
  • No
  • No
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together