Order Entry
United States
ContactUsLinkComponent
 
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SK3]
 
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SK3]
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 10002-308
  • 10002-312
  • 10002-310
  • Antigen Symbol:
  • CD4
  • CD4
  • CD4
  • Antigen Name:
  • CD4 molecule
  • CD4 molecule
  • CD4 molecule
  • Antigen Synonyms:
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • SK3
  • SK3
  • SK3
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 920
  • 920
  • 920
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Cross Adsorption:
  • No
  • No
  • No
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.

 

Frequently Bought Together