About this item
The L243 antibody reacts with a member of the human MHC Class II antigens, HLA-DR. The HLA-DR antigen is expressed on B lymphocytes, activated T lymphocytes, activated NK cells, monocytes, macrophages, other antigen presenting cells and progenitor cells. The L243 antibody is specific to an epitope on the alpha subunit of the heterodimeric HLA-DR protein and binds a different epitope than the LN3 antibody clone. It does not cross-react with HLA-DP or HLA-DQ. This antibody is reported to be cross-reactive with non-human primates including Chimpanzee, Cynomolgus, Rhesus and Baboon. Please choose the appropriate format for each application.
Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: L243
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Specifications
- Catalog No:
- 10002-306
- 10002-304
- 10002-302
- Antigen Symbol:
- HLA-DRA
- HLA-DRA
- HLA-DRA
- Antigen Name:
- Major histocompatibility Complex, class II, DR alpha
- Major histocompatibility Complex, class II, DR alpha
- Major histocompatibility Complex, class II, DR alpha
- Antigen Synonyms:
- MLRW|HLA-DRA1
- MLRW|HLA-DRA1
- MLRW|HLA-DRA1
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- L243
- L243
- L243
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 3122
- 3122
- 3122
- Isotype:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa
- Flow Cytometry:
- Yes
- Yes
- Yes
- Cross Adsorption:
- No
- No
- No
- Storage Temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.