Order Entry
United States
ContactUsLinkComponent
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MY31]
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MY31]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MY31]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10753-854
  • 10753-856
  • Antigen Symbol:
  • NCAM1
  • NCAM1
  • Antigen Name:
  • Neural Cell Adhesion Molecule 1
  • Neural Cell Adhesion Molecule 1
  • Antigen Synonyms:
  • NCAM|MSK39|CD56
  • NCAM|MSK39|CD56
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • MY31
  • MY31
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4684
  • 4684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Cross Adsorption:
  • No
  • No
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together