Order Entry
United States
ContactUsLinkComponent
Rat IgG2b Isotype Control [clone: LTF-2]
Rat IgG2b Isotype Control [clone: LTF-2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Rat IgG2b Isotype Control [clone: LTF-2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10141-172
  • 10753-770
  • Antigen Symbol:
  • LTF-2
  • IgG2 ß
  • Antigen Name:
  • Immunoglobulin G2 beta
  • Antibody Type:
  • Control
  • Control
  • Clonality:
  • Monoclonal
  • Clone:
  • LTF-2
  • LTF-2
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2b
  • IgG2b
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Cross Adsorption:
  • No
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store undiluted at 2-8°C, do not freeze.
  • Tested Applications:
  • Functional Assays, Immunofluorescence Microscopy

Specifications

Related Information

Frequently Bought Together