Order Entry
United States
ContactUsLinkComponent
Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]
Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10753-762
  • 10753-760
  • 10753-764
  • Antigen Symbol:
  • MS4A1
  • MS4A1
  • MS4A1
  • Antigen Name:
  • Membrane-Spanning 4-Domains, Subfamily A, Member 1
  • Membrane-Spanning 4-Domains, Subfamily A, Member 1
  • Membrane-Spanning 4-Domains, Subfamily A, Member 1
  • Antigen Synonyms:
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 2H7
  • 2H7
  • 2H7
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 931
  • 931
  • 931
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Cross Adsorption:
  • No
  • No
  • No
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together