Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L17F12]
Supplier: Tonbo Biosciences
Certificates
About this item
The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Specifications
- Catalog No:
- 10753-754
- 10753-758
- 10753-756
- Antigen Symbol:
- CD5
- CD5
- CD5
- Antigen Name:
- CD5 molecule
- CD5 molecule
- CD5 molecule
- Antigen Synonyms:
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- L17F12
- L17F12
- L17F12
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 921
- 921
- 921
- Isotype:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa
- Flow Cytometry:
- Yes
- Yes
- Yes
- Cross Adsorption:
- No
- No
- No
- Storage Temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
Specifications
Related Information
Frequently Bought Together