Order Entry
United States
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-2.10]
Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-2.10]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-2.10]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10753-742
  • 10753-746
  • 10753-744
  • Antigen Symbol:
  • CD2
  • CD2
  • CD2
  • Antigen Name:
  • CD2 molecule
  • CD2 molecule
  • CD2 molecule
  • Antigen Synonyms:
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • RPA-2.10
  • RPA-2.10
  • RPA-2.10
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 914
  • 914
  • 914
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Cross Adsorption:
  • No
  • No
  • No
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together