Sodium phosphate dibasic, anhydrous USP, Ph. Eur. endotoxin tested
Supplier: Avantor
Certificates
About this item
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
CAS Number:7558-79-4
:
Formula:Na₂HPO₄
MDL Number:MFCD00003496
Molecular Weight:141.96 g/mol
Melting Pt:250
Merck Index:12,08805
Storage Temperature:Ambient
EINECS:231-448-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance of Solution:PASS / Fail
- Arsenic (2.4.2, Method A):≤ 2 ppm
- Chlorides:≤ 200 ppm
- Endotoxin:0.5000 EU/g
- Heavy Metals (2.4.8):≤ 10 ppm
- Identification A:PASS / Fail
- Identification B:PASS / Fail
- Identification C:PASS / Fail
- Identification D:PASS / Fail
- Iron:≤ 20 ppm
- Loss on Drying (2.2.32):≤ 1.0%
- Monosodium phosphate:≤ 2.5%
- Purity (dried substance):98.0 - 101.0%
- Reducing Substances:PASS / Fail
- Sulfates:≤ 500 ppm
- Additional Information, country of origin:DE
- Additional information:Material is of non-animal, non-human origin
- Additional information, storage conditions:15ºC to 30ºC
Specification Test Results
Related Information
Certificates
Search by Lot NumberAT-SOD PHOS DIBASIC ANH_0404_Certificate of SpecificationsREV2.pdf spec_VWRB0404-12KG
Product specification
ghs_VWRB0404
SDS
Production Insight Solutions for Controlled Environments and Manufacturing Processes Production insight Solutions for Critical Environments and Manufacturing Processes Production insight v1 final1 VWR Bioprocessing Chemical and Excipients VWR Bioprocessing Chemicals and Excipients
Product Literature
More
VWRB0404-12KG_BSE
BSE/TSE Certificates
Frequently Bought Together