ContactUsLinkComponent
AC/DC Magnetic Meter/Datalogger, Extech
AC/DC Magnetic Meter/Datalogger, Extech
Catalog # 10017-256
Supplier:  Avantor
UL Listed
undefined
AC/DC Magnetic Meter/Datalogger, Extech
Catalog # 10017-256
Supplier:  Avantor
UL Listed
Specifications
  • Accuracy:
    ±(5% + 20d), ±(5% + 10d)
  • Measurement Range:
    AC: 150mT/150G, DC: 300mT/300G
  • Description:
    AC/DC Magnetic Meter/Datalogger
  • Environmentally Preferable:
  • Output:
    RS-232
  • Cat. No.:
    10017-256
  • Supplier no.:
    SDL900
Specifications
About this item
The AC/DC Magnetic Meter/Datalogger measures and datalogs both AC and DC magnetic field in Gauss and mT (milli Tesla) with a uni-axial Hall effect sensor and built-in ATC
  • Adjustable data sampling rate from 1 to 3600 seconds
  • Memory stores 99 readings manually
  • Standard features include N pole/S pole indicator
  • Zero button for DC measurement
  • Data hold and min./max.
  • Auto power off with disable
  • RS-232 interface
Green/orange housing.
Certifications: CE compliant.
Ordering information: Supplied complete with uni-axial magnetic probe sensor with protective cover, SD memory card, universal AC adaptor with multi-plugs (US, EU, UK, AUS), 6 AA batteries, and hard carrying case.