Disposable Ultrafilter Units, Advantec MFS
Supplier: Advantec MFS
Certificates
About this item
Advantec's USY series disposable ultrafiltration units offers maximum sample recovery with a low absorption polysulfone membrane.
- MWC: 10, 50, 200 kDa
- Easy to use: simple operation requires only syringe pressurization
- Concentrate samples <2 ml with built-in polysulfone membrane
- Clear housing enables visual confirmation of filtration process
Operation: Dispense sample into retentive cell by pipet. Secure cap. Using a syringe, pressurize the unit with air. When filtration is complete, disassemble unit and concentrated sample/filtrate can be analyzed.
Ordering information: Syringe not included.
Delivery: Package includes 24 retentive cells, 24 filtrate cells, and 4 reusable cell caps.
Specifications
- Catalog No:
- 101104-966
- 101104-964
- 101104-968
- Filtration Area:
- 2.0 cm²
- 2.0 cm²
- 2.0 cm²
- Capacity:
- 2 mL (Cell Capacity)
- 2 mL (Cell Capacity)
- 2 mL (Cell Capacity)
- Housing Material:
- Cells: Acrylic, Cap: Polypropylene
- Cells: Acrylic, Cap: Polypropylene
- Cells: Acrylic, Cap: Polypropylene
- Sterilization:
- 25% Ethanol or 5% Formalin
- 25% Ethanol or 5% Formalin
- 25% Ethanol or 5% Formalin
- Max. Pressure:
- 0.29 Mpa (42 psi)
- 0.29 Mpa (42 psi)
- 0.29 Mpa (42 psi)
- Max. Temperature:
- 50°C (122°F)
- 50°C (122°F)
- 50°C (122°F)
- Min. Sample Volume:
- 0.5 mL
- 0.5 mL
- 0.5 mL
- Ø:
- 16 mm
- 16 mm
- 16 mm
Specifications
Frequently Bought Together