Specifications
- Description:Stainless Steel Stitch Cutter
- Length:8.9 cm (3.5")
- Cat. No.:10193-782
- Supplier no.:06-3030
Specifications
About this item
The Stitch Cutter is used for general suture removal
Appearing similar to a scalpel, this instrument requires no handle and is a simple and convenient tool for removing basic sutures. The slightly curved, flat, bull nose tip slides under the suture and when rotated, gently removes the suture from the site. This product is the "midi" version and measures approximately 8.9cm (3.5") overall. Supplied sterile, in boxes of 100.