Specifications
- Sterility:Non sterile
- Material:Stainless Steel/Plastic
- Cat. No.:95045-498
- Blade no.:#11
- Supplier no.:06-3132
Specifications
About this item
The SklarSafe™ Safety Scalpel features a protective, transparent sheath that can be opened, closed, and locked with a single hand
The sturdy "sure-grip" handle provides an audible click when the sheath is advanced to a locked position. This instrument is OSHA compliant and individually sealed in sturdy film and Tyvek packaging. Nonsterile, disposable scalpels are supplied in a box of 10.