Order Entry
United States
ContactUsLinkComponent
Anti-SYN Rabbit Polyclonal Antibody
Anti-SYN Rabbit Polyclonal Antibody
Catalog # 10007-978
Supplier:  BioVision
undefined
Anti-SYN Rabbit Polyclonal Antibody
Catalog # 10007-978
Supplier:  BioVision
Supplier Number:  AB18814-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Synapsin
  • Antigen Symbol:
    SYN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    6853
  • Antigen Synonyms:
    SYN|syn
  • Storage Buffer:
    PBS, pH 7.2, containing 30% glycerol, 0.5% BSA, and 0.01% thimerosal.
  • Storage Temperature:
    -20 °C
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    4°C
  • Immunogen:
    Synthetic peptide corresponding to residues surrounding amino acid 12 of human synapsin
  • Purification:
    Affinity purified
  • Cat. No.:
    10007-978
  • Supplier no.:
    6008-100

Specifications

About this item

Synapsins are abundant brain proteins essential for regulating neurotransmitter release. All synapsins contain a short N-terminal domain that is highly conserved and phosphorylated by PKA or CaM kinase I. Phosphorylation of synapsin N-terminal domain at Ser9 inhibits its binding to phospholipids and diassociates synapsins from synaptic vesicles.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat