Chemical-Resistant Labels, Diversified Biotech, Tough-Tags®
Supplier: Diversified Biotech
Certificates
About this item
Tough-Tags® are chemically resistant, flexible, polyester labels that can withstand autoclaving, boiling water baths, freezing to –40 °C, or heating up to 121°C without degrading. The thermal transfer Tough-Tags can withstand up to 150 °C.
They adhere strongly to plastics and other materials. Tough-Tags® are thin and will not bind centrifuge rotors.
Frequently Bought Together