TubePopper, Diversified Biotech
Catalog # 10141-898
Supplier: Diversified Biotech
Specifications
- Description:TubePopper
- Cat. No.:10141-898
- Supplier no.:TPOP-1000
Specifications
About this item
For one-handed opening and closing of microtubes
- “Pry open” design means tubes pop right open, saving your fingers from the repetitive stress of opening microtubes
- Bottom portion of TubePopper allows caps to be pressed closed without snagging glove fingers
Works with most flat-top and domed microtubes. TubePopper can be worn on any finger so you can find the perfect pipetting fit.
Ring design allows researcher to wear the TubePopper on the pipetting hand.