Breathe-Easier Sealing Film, Diversified Biotech
10141-844
- L×W:152.4×82.55 mm (6×3.23")
- Cat. No.:10141-844
- Supplier no.:BERM-2000
Allows rapid and uniform transmission of oxygen, carbon dioxide and water vapors to stimulate cell growth.
- Non-cytotoxic and sterile - Membranes are free of cytotoxins and also gamma irradiated to assure sterility
- Chemically and thermally stable - Resists a majority of caustic agents and organic solvents as well as exposure to temperatures from -20°C to 80°C
- Easy sample retrieval - membrane is easily pierced by pipet tips
- US Patent # 5,858,770