ContactUsLinkComponent
Kitchen Chemistry
Kitchen Chemistry
Catalog # 470200-260
Supplier:  Lab-Aids
undefined
Kitchen Chemistry
Catalog # 470200-260
Supplier:  Lab-Aids
Supplier Number:  190
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Kitchen Chemistry
  • No. of lab groups:
    12
  • Cat. No.:
    470200-260
  • Includes:
    Teacher Guide, SDS, Student Guide/Worksheets
  • Supplier no.:
    190
Specifications
About this item
How Do Common Household Chemicals React?
  • Emphasize experimental design and observation
  • Develop scientific reasoning skills
  • Kit requires one ~50 minute class period to complete
The primary purpose of this Lab-Aid is not to identify the common kitchen chemicals, but to observe how they react. Students first explore some physical and chemical properties of several common kitchen substances (four powders). Then, they determine some properties of the powders by using three liquid indicators. After determining the physical and chemical properties, students attempt to identify some of the mystery powders provided using the results from their experimental observation. The solutions and powders necessary for up to 50 students to complete the activities are included in this kit.
Ordering information: Kit includes Chemplates® with spatula, Droppers, Bottles of Powders 1-4, Bottles of Indicator M, Vials of "unknown" 1-4, Drop control bottle of Indicator A, and Drop control bottle of Indicator C.