Order Entry
United States
ContactUsLinkComponent
ICP-MS Tuning Standard solution for 200.8, Specpure®, Thermo Scientific
ICP-MS Tuning Standard solution for 200.8, Specpure®, Thermo Scientific
Catalog # AA45641-AE
CAS Number:  
ICP-MS Tuning Standard solution for 200.8, Specpure®, Thermo Scientific
Catalog # AA45641-AE
Supplier Number:  045641-AE
CAS Number:  

Specifications

  • Size:
    100 ml
  • MDL No.:
    MFCD00240453
  • Cat. No.:
    AA45641-AE
  • Supplier no.:
    45641-AE

Specifications

About this item

Liquid

Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00240453