Yttrium, Plasma Standard Solution, Specpure®, Y 10 µg/ml, Thermo Scientific
Catalog # AA45268-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Size:100 ml
- MDL No.:MFCD00011468
- Cat. No.:AA45268-AE
- Element:Yttrium
- Supplier no.:45268-AE
Specifications
About this item
Liquid