Manganese, plasma standard solution, Specpure®, Mn 10µg/ml, Thermo Scientific
Catalog # AA45258-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Size:100 ml
- MDL No.:MFCD00011111
- Cat. No.:AA45258-AE
- Element:Manganese
- Supplier no.:45258-AE
Specifications
About this item
Liquid
Molecular Formula: Mn in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111