Combustion Boats, Molybdenum, Thermo Scientific
Supplier: Avantor
Certificates
About this item
These boats are made up of molybdenum
- High melting point
- High resistance to corrosion
Used for aperture cleaning and vacuum evaporation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.