Order Entry
United States
ContactUsLinkComponent
Imidazole, 0.2M Buffer Soln., pH 7.0, Low Endotoxin, Thermo Scientific
Imidazole, 0.2M Buffer Soln., pH 7.0, Low Endotoxin, Thermo Scientific
Certificates
Imidazole, 0.2M Buffer Soln., pH 7.0, Low Endotoxin, Thermo Scientific

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    288-32-4
  • MDL No.:
    MFCD00005183
  • Storage Temperature:
    2°C to 8°C
  • Physical Form:
    Liquid
  • Appearance:
    Colorless

Specifications

Related Information

Frequently Bought Together