Order Entry
United States
ContactUsLinkComponent
Mitomycin C
Toxic
Health
Mitomycin C
Catalog # AAJ63193-MA
CAS Number:  
Toxic
Health
Mitomycin C
Catalog # AAJ63193-MA
Supplier Number:  J63193-MA
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 mg
  • Cat. No.:
    AAJ63193-MA
  • Supplier no.:
    J63193-MA

Specifications

About this item

 :
Formula:C₁₅H₁₈N₄O₅
MDL Number:MFCD00078109
 :334.33 g/mol
Merck Index:14,6215
Melting Pt:360 °C
Storage Temperature:Ambient
EINECS:200-008-6
UN:2811
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:50-07-7