Order Entry
United States
Orders LinkContactUsLinkComponent
 

4-methoxy-1H-indazol-3-amine 97%

4-methoxy-1H-indazol-3-amine 97%
Supplier:  
Certificates

0 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 886362-07-8 Molecular Formula: C8H9N3O Formula Weight: 163.18 Melting Point: 115-118° MDL No.: MFCD08276946

Synonyms:  3-Amino-4-methoxy-1H-indazole, 4-Methoxy-1H-indazol-3-amine
  • CAS Number:886362-07-8
  • Formula:C₈H₉N₃O
  • MDL Number:MFCD08276946
  • Molecular Weight:163.18 g/mol
  • Melting Pt:115…118 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Assay Minimum:
    97
Product Family Options