- Description:O-Ring for AC 10, Nitrile Rubber
- Dimensions:6.75 x 1.78 mm
- Cat. No.:101724-990
- Supplier no.:19504301
Resistant to aqueous solutions and solvents used in liquid chromatography.
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.