About this item
The ExoProStar™ S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar™ S improves digestion efficiency with no degradation of the target PCR product. Sample remain completely intact and ready for immediate use in manual or automated processes. For maximum flexibility the enzymes are supplied in two separate tubes.
- Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
- Enzymes provided in two separate tubes - just two simple pipetting steps are needed to prepare the reaction
- Fast 15 minute protocol
- Scalable for different reaction sizes
- No loss of PCR product
- Easy to automate
- Complete heat inactivation of the enzymes within 10 minutes
Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction.
The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Next-Generation Sequencing (NGS) Solutions