Order Entry
United States
Orders LinkContactUsLinkComponent
Multi-Element Qc-19 Check Standard Solution

Multi-Element Qc-19 Check Standard Solution

Multi-Element Qc-19 Check Standard Solution
Supplier:  BeanTown Chemical
Certificates

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BT224270-100ML
  • BT224270-500ML
  • MDL No.:
  • MFCD00210655
  • MFCD00210655
  • UN No.:
  • 3264
  • 3264
  • GHS signal:
  • Danger
  • Danger

Specifications

Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 100 ml
    HDPE plastic bottle
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    BT224270-100ML
    MDL No.
    MFCD00210655
    UN No.
    3264
    GHS signal
    Danger
  • 500 ml
    HDPE plastic bottle
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $683.00
    $683.00
    Each (500ml)
       $683.00

    Supplier Number: 224270-500ML
    Specifications:
    More
    Specifications
    Cat. No.
    BT224270-500ML
    MDL No.
    MFCD00210655
    UN No.
    3264
    GHS signal
    Danger

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.