Order Entry
United States
ContactUsLinkComponent
Mercury, Plasma Standard Solution, Hg 1000 µg/ml
Mercury, Plasma Standard Solution, Hg 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Mercury, Plasma Standard Solution, Hg 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00011035
  • UN No.:
    3264
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together