Tetraethylammonium hydroxide 35% (w/w) in water
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:77-98-5
:
Formula:C₈H₂₁NO
MDL Number:MFCD00009024
:147.26
Density:1.023 g/mL
EINECS:201-073-3
UN:3267
ADR:8,II
Frequently Bought Together