Order Entry
United States
ContactUsLinkComponent
Cadmium, Plasma Standard Solution, Cd 1000 µg/ml
Cadmium, Plasma Standard Solution, Cd 1000 µg/ml
Supplier:  BeanTown Chemical
Certificates
Cadmium, Plasma Standard Solution, Cd 1000 µg/ml
Supplier:  BeanTown Chemical

Specifications

  • MDL No.:
    MFCD00010914
  • UN No.:
    3264
  • GHS signal:
    Danger

Specifications

Related Information

Frequently Bought Together